The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300743.1 | NP_001287672.1 | 13 - 14 | 1390 | 72 |
NM_020314.5 | NP_064710.4 | 15 - 16 | 1591 | 72 | |
XM_005255435.3 | XP_005255492.1 | 15 - 16 | 1543 | 72 | |
XM_006721065.3 | XP_006721128.1 | 13 - 14 | 1342 | 72 | |
XM_011545896.2 | XP_011544198.1 | 15 - 16 | 1292 | 72 | |
XM_011545897.2 | XP_011544199.1 | 8 - 9 | 659 | 72 | |
GenBank mRNA | AF461052.2 | - | 9 - 10 | 1087 | 72 |
AK024693.1 | - | 11 - 12 | 861 | 72 | |
AK095078.1 | - | 13 - 14 | 1087 | 72 | |
AK290286.1 | - | 15 - 16 | 1288 | 72 | |
AL136744.1 | - | 2 - 3 | 204 | 72 | |
AL832159.1 | - | 15 - 16 | 1287 | 72 | |
BC014900.2 | - | 14 - 15 | 1259 | 72 | |
BC050464.1 | - | 15 - 16 | 1331 | 72 | |
BC058845.1 | - | 15 - 16 | 1290 | 72 | |
BX537867.1 | - | 15 - 16 | 1303 | 72 |