The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317813.1 | NP_001304742.1 | 3 - 4 | 771 | 68 |
NM_001317814.1 | NP_001304743.1 | 3 - 4 | 718 | 68 | |
NM_016612.3 | NP_057696.2 | 2 - 3 | 647 | 68 | |
XM_006716352.3 | XP_006716415.1 | 2 - 3 | 251 | 68 | |
XM_011544550.2 | XP_011542852.1 | 2 - 3 | 251 | 68 | |
XM_011544553.2 | XP_011542855.1 | 3 - 4 | 1483 | 68 | |
XM_017013532.1 | XP_016869021.1 | 2 - 3 | 1322 | 68 | |
GenBank mRNA | AF223466.1 | - | 2 - 3 | 483 | 68 |
BC132799.1 | - | 2 - 3 | 511 | 68 | |
BC132801.1 | - | 2 - 3 | 511 | 68 | |
BM552840.1 | - | 3 - 4 | 653 | 68 | |
BM926580.1 | - | 2 - 3 | 490 | 68 | |
BU166030.1 | - | 3 - 4 | 691 | 68 |