Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
Chromosome Location
Chr.2: 73385758 - 73609919 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015120.4 NP_055935.4 2 - 3 561 114
GenBank mRNA AJ417593.2 - 2 - 3 561 114

View Full Product Details