The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015120.4 | NP_055935.4 | 4 - 5 | 873 | 121 |
GenBank mRNA | AJ417593.2 | - | 4 - 5 | 873 | 121 |
AX751581.1 | - | 3 - 4 | 747 | 121 |