The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006662.2 | NP_006653.2 | 5 - 6 | 874 | 63 |
GenBank mRNA | AB621816.1 | - | 5 - 6 | 941 | 63 |
AF143946.1 | - | 1 - 2 | 108 | 63 | |
AK128030.1 | - | 5 - 6 | 874 | 63 | |
BC025284.1 | - | 2 - 3 | 351 | 63 | |
BC063578.1 | - | 3 - 4 | 668 | 63 |