The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006662.2 | NP_006653.2 | 29 - 30 | 6881 | 92 |
GenBank mRNA | AF143946.1 | - | 24 - 25 | 5929 | 92 |
AK128030.1 | - | 29 - 30 | 6881 | 92 |