The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243186.1 | NP_001230115.1 | 2 - 3 | 620 | 76 |
NM_002648.3 | NP_002639.1 | 2 - 3 | 620 | 76 | |
GenBank mRNA | AK301158.1 | - | 2 - 3 | 178 | 76 |
BC020224.1 | - | 2 - 3 | 553 | 76 | |
DA481890.1 | - | 2 - 3 | 513 | 76 | |
DQ022562.1 | - | 2 - 3 | 689 | 76 | |
M16750.1 | - | 2 - 3 | 212 | 76 | |
M24779.1 | - | 2 - 3 | 478 | 76 | |
M54915.1 | - | 2 - 3 | 540 | 76 |