The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243186.1 | NP_001230115.1 | 5 - 6 | 1213 | 106 |
NM_002648.3 | NP_002639.1 | 5 - 6 | 1213 | 106 | |
GenBank mRNA | AK301158.1 | - | 5 - 6 | 771 | 106 |
AK301314.1 | - | 2 - 3 | 514 | 106 | |
BC020224.1 | - | 5 - 6 | 1146 | 106 | |
DQ022562.1 | - | 5 - 6 | 1282 | 106 | |
M16750.1 | - | 5 - 6 | 805 | 106 | |
M24779.1 | - | 5 - 6 | 1071 | 106 | |
M54915.1 | - | 5 - 6 | 1133 | 106 |