The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007077.4 | NP_009008.2 | 5 - 6 | 754 | 73 |
XM_005267293.4 | XP_005267350.1 | 6 - 7 | 1197 | 73 | |
XM_011536371.2 | XP_011534673.1 | 6 - 7 | 767 | 73 | |
XM_011536372.2 | XP_011534674.1 | 5 - 6 | 1161 | 73 | |
GenBank mRNA | BC001259.1 | - | 5 - 6 | 537 | 73 |
BT006701.1 | - | 4 - 5 | 365 | 73 |