Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIT2
Entrez Gene ID
9815
Gene Name
GIT ArfGAP 2
Gene Aliases
CAT-2, CAT2, PKL
Chromosome Location
Chr.12: 109929792 - 109996389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135213.1 NP_001128685.1 1 - 2 213 87
NM_001135214.1 NP_001128686.1 1 - 2 213 87
NM_014776.3 NP_055591.2 1 - 2 213 87
NM_057169.3 NP_476510.1 1 - 2 213 87
NM_057170.3 NP_476511.1 1 - 2 213 87
NM_139201.2 NP_631940.2 1 - 2 213 87
XM_005253997.3 XP_005254054.1 1 - 2 208 87
XM_005254000.3 XP_005254057.1 1 - 2 208 87
XM_006719707.3 XP_006719770.1 1 - 2 208 87
XM_006719708.3 XP_006719771.1 1 - 2 208 87
XM_006719709.3 XP_006719772.1 1 - 2 208 87
XM_006719712.3 XP_006719775.1 1 - 2 208 87
XM_006719713.3 XP_006719776.1 1 - 2 205 87
XM_006719714.3 XP_006719777.1 1 - 2 208 87
XM_017020258.1 XP_016875747.1 1 - 2 208 87
XM_017020259.1 XP_016875748.1 1 - 2 205 87
XM_017020260.1 XP_016875749.1 1 - 2 205 87
XM_017020261.1 XP_016875750.1 1 - 2 203 87
GenBank mRNA AB451294.1 - 1 - 2 48 87
AB451428.1 - 1 - 2 48 87
AF124491.1 - 1 - 2 48 87
AK222736.1 - 1 - 2 120 87
AK304240.1 - 1 - 2 197 87
AK308304.1 - 1 - 2 128 87
BC001379.2 - 1 - 2 113 87
BP871813.1 - 1 - 2 213 87
BT007312.1 - 1 - 2 48 87
CR533568.1 - 1 - 2 48 87
D63482.2 - 1 - 2 119 87

View Full Product Details