Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GIT2
- Entrez Gene ID
-
9815
- Gene Name
- GIT ArfGAP 2
- Gene Aliases
-
CAT-2, CAT2, PKL
- Chromosome Location
-
Chr.12: 109929792 - 109996389 on Build GRCh38
View Full Product Details