Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLTP
Entrez Gene ID
5360
Gene Name
phospholipid transfer protein
Gene Aliases
BPIFE, HDLCQ9
Chromosome Location
Chr.20: 45898620 - 45912364 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242920.1 NP_001229849.1 7 - 8 899 64
NM_001242921.1 NP_001229850.1 7 - 8 811 64
NM_006227.3 NP_006218.1 9 - 10 1184 64
NM_182676.2 NP_872617.1 8 - 9 1028 64
GenBank mRNA AB076694.1 - 9 - 10 968 64
AF052157.1 - 6 - 7 686 64
AK092397.1 - 9 - 10 925 64
AK092523.1 - 8 - 9 816 64
AK097009.1 - 8 - 9 1482 64
AK222690.1 - 9 - 10 972 64
AK289371.1 - 8 - 9 816 64
AK293150.1 - 7 - 8 727 64
AK299181.1 - 7 - 8 691 64
BC005045.1 - 8 - 9 809 64
BC019847.1 - 9 - 10 966 64
BC019898.1 - 9 - 10 959 64
L26232.1 - 9 - 10 974 64

View Full Product Details