The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206855.1 | NP_001193784.1 | 5 - 6 | 726 | 79 |
NM_003227.3 | NP_003218.2 | 8 - 9 | 1146 | 79 | |
XM_005250553.4 | XP_005250610.1 | 9 - 10 | 1452 | 79 | |
XM_017012573.1 | XP_016868062.1 | 9 - 10 | 1254 | 79 | |
GenBank mRNA | AF067864.1 | - | 8 - 9 | 1132 | 79 |
BC142630.1 | - | 8 - 9 | 1132 | 79 | |
BX384750.2 | - | 5 - 6 | 726 | 79 |