The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 14 - 15 | 14385 | 140 |
NM_033026.5 | NP_149015.2 | 14 - 15 | 14385 | 140 | |
XM_017012006.1 | XP_016867495.1 | 12 - 13 | 8565 | 140 | |
XM_017012007.1 | XP_016867496.1 | 12 - 13 | 8549 | 140 | |
GenBank mRNA | AB011131.1 | - | 8 - 9 | 2929 | 140 |
BC001304.1 | - | 6 - 7 | 618 | 140 | |
BC108289.1 | - | 5 - 6 | 587 | 140 |