The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 15 - 16 | 14513 | 95 |
NM_033026.5 | NP_149015.2 | 15 - 16 | 14513 | 95 | |
XM_017012006.1 | XP_016867495.1 | 13 - 14 | 8693 | 95 | |
GenBank mRNA | AB011131.1 | - | 9 - 10 | 3057 | 95 |