The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 16 - 17 | 14537 | 138 |
NM_033026.5 | NP_149015.2 | 16 - 17 | 14537 | 138 | |
XM_017012006.1 | XP_016867495.1 | 14 - 15 | 8717 | 138 | |
GenBank mRNA | AB011131.1 | - | 10 - 11 | 3081 | 138 |