The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 12 - 13 | 14118 | 123 |
NM_033026.5 | NP_149015.2 | 12 - 13 | 14118 | 123 | |
XM_017012006.1 | XP_016867495.1 | 10 - 11 | 8298 | 123 | |
XM_017012007.1 | XP_016867496.1 | 10 - 11 | 8282 | 123 | |
GenBank mRNA | AB011131.1 | - | 6 - 7 | 2662 | 123 |
BC108289.1 | - | 3 - 4 | 320 | 123 |