Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNW1
Entrez Gene ID
22938
Gene Name
SNW domain containing 1
Gene Aliases
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP
Chromosome Location
Chr.14: 77717599 - 77761220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012245.2 NP_036377.1 13 - 14 1435 97
XM_005267413.3 XP_005267470.1 12 - 13 1404 97
GenBank mRNA AF045184.1 - 13 - 14 1435 97
AK292274.1 - 13 - 14 1434 97
AK302246.1 - 13 - 14 1306 97
AK316087.1 - 13 - 14 1466 97
AX775981.1 - 13 - 14 1429 97
BC040112.2 - 12 - 13 1395 97
BC046105.2 - 12 - 13 1395 97
BC108903.1 - 13 - 14 1428 97
BT020060.1 - 12 - 13 1408 97
BT020061.1 - 12 - 13 1408 97
CR457433.1 - 12 - 13 1408 97
U51432.1 - 13 - 14 1429 97

View Full Product Details