The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318844.1 | NP_001305773.1 | 7 - 8 | 735 | 69 |
NM_012245.2 | NP_036377.1 | 7 - 8 | 735 | 69 | |
XM_005267413.3 | XP_005267470.1 | 7 - 8 | 801 | 69 | |
GenBank mRNA | AF045184.1 | - | 7 - 8 | 735 | 69 |
AK292274.1 | - | 7 - 8 | 734 | 69 | |
AK293619.1 | - | 7 - 8 | 737 | 69 | |
AK302246.1 | - | 7 - 8 | 606 | 69 | |
AK316087.1 | - | 7 - 8 | 766 | 69 | |
AX775981.1 | - | 7 - 8 | 729 | 69 | |
BC032377.2 | - | 6 - 7 | 695 | 69 | |
BC040112.2 | - | 6 - 7 | 695 | 69 | |
BC046105.2 | - | 6 - 7 | 695 | 69 | |
BC065286.1 | - | 6 - 7 | 713 | 69 | |
BC105585.1 | - | 7 - 8 | 733 | 69 | |
BC108903.1 | - | 7 - 8 | 728 | 69 | |
BT020060.1 | - | 6 - 7 | 708 | 69 | |
BT020061.1 | - | 6 - 7 | 708 | 69 | |
U51432.1 | - | 7 - 8 | 729 | 69 |