The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318844.1 | NP_001305773.1 | 1 - 2 | 39 | 69 |
NM_012245.2 | NP_036377.1 | 1 - 2 | 39 | 69 | |
XM_005267413.3 | XP_005267470.1 | 1 - 2 | 105 | 69 | |
GenBank mRNA | AF045184.1 | - | 1 - 2 | 39 | 69 |
AK292274.1 | - | 1 - 2 | 38 | 69 | |
AK293619.1 | - | 1 - 2 | 41 | 69 | |
BC105585.1 | - | 1 - 2 | 37 | 69 | |
BC108903.1 | - | 1 - 2 | 32 | 69 |