Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM1
Entrez Gene ID
2911
Gene Name
glutamate metabotropic receptor 1
Gene Aliases
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13
Chromosome Location
Chr.6: 146027634 - 146437598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278064.1 NP_001264993.1 2 - 3 1268 78
NM_001278065.1 NP_001264994.1 3 - 4 1315 78
NM_001278066.1 NP_001264995.1 2 - 3 1268 78
NM_001278067.1 NP_001264996.1 2 - 3 1268 78
XM_011535782.1 XP_011534084.1 3 - 4 1418 78
XM_017010783.1 XP_016866272.1 3 - 4 1427 78
XM_017010784.1 XP_016866273.1 3 - 4 1463 78
XM_017010785.1 XP_016866274.1 3 - 4 1423 78
XM_017010786.1 XP_016866275.1 3 - 4 1423 78
XM_017010787.1 XP_016866276.1 3 - 4 1459 78
GenBank mRNA AB208837.1 - 2 - 3 935 78
BC136280.1 - 2 - 3 980 78
BC143777.1 - 2 - 3 980 78
BC143779.1 - 2 - 3 978 78
L76627.1 - 2 - 3 948 78
L76631.1 - 3 - 4 1318 78
U31215.1 - 2 - 3 1183 78
U31216.1 - 2 - 3 948 78

View Full Product Details