The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304799.1 | NP_001291728.1 | 4 - 5 | 444 | 77 |
NM_022493.2 | NP_071938.1 | 3 - 4 | 362 | 77 | |
GenBank mRNA | AK025641.1 | - | 3 - 4 | 321 | 77 |
AK025861.1 | - | 1 - 2 | 61 | 77 | |
AK056467.1 | - | 2 - 3 | 1439 | 77 | |
AK095675.1 | - | 1 - 2 | 707 | 77 | |
AK223005.1 | - | 3 - 4 | 321 | 77 | |
AK301023.1 | - | 3 - 4 | 321 | 77 | |
AY129231.1 | - | 3 - 4 | 315 | 77 | |
BC030248.1 | - | 3 - 4 | 314 | 77 | |
DA153375.1 | - | 3 - 4 | 362 | 77 | |
DB126434.1 | - | 3 - 4 | 340 | 77 |