The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022773.2 | NP_073610.2 | 5 - 6 | 739 | 81 |
NR_033645.1 | 6 - 7 | 868 | 81 | ||
NR_033646.1 | 7 - 8 | 1046 | 81 | ||
XM_006720928.2 | XP_006720991.1 | 5 - 6 | 1012 | 81 | |
XM_006720929.3 | XP_006720992.1 | 6 - 7 | 893 | 81 | |
XM_011522614.2 | XP_011520916.1 | 5 - 6 | 737 | 81 | |
XM_011522617.2 | XP_011520919.1 | 3 - 4 | 388 | 81 | |
XM_011522618.2 | XP_011520920.1 | 5 - 6 | 737 | 81 | |
XM_017023575.1 | XP_016879064.1 | 5 - 6 | 737 | 81 | |
XM_017023576.1 | XP_016879065.1 | 4 - 5 | 503 | 81 | |
XM_017023577.1 | XP_016879066.1 | 6 - 7 | 1168 | 81 | |
XM_017023578.1 | XP_016879067.1 | 4 - 5 | 588 | 81 | |
XM_017023579.1 | XP_016879068.1 | 4 - 5 | 596 | 81 | |
XM_017023580.1 | XP_016879069.1 | 7 - 8 | 1318 | 81 | |
GenBank mRNA | AB075873.1 | - | 5 - 6 | 723 | 81 |
AK022743.1 | - | 5 - 6 | 737 | 81 | |
AK025955.1 | - | 5 - 6 | 671 | 81 | |
AK093050.1 | - | 2 - 3 | 246 | 81 | |
AK125201.1 | - | 6 - 7 | 870 | 81 | |
AK225065.1 | - | 5 - 6 | 737 | 81 | |
AK225406.1 | - | 5 - 6 | 724 | 81 | |
AK294329.1 | - | 7 - 8 | 1044 | 81 | |
BC010738.1 | - | 4 - 5 | 504 | 81 |