The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022773.2 | NP_073610.2 | 6 - 7 | 906 | 144 |
NR_033645.1 | 7 - 8 | 1035 | 144 | ||
NR_033646.1 | 8 - 9 | 1213 | 144 | ||
XM_006720928.2 | XP_006720991.1 | 6 - 7 | 1179 | 144 | |
XM_006720929.3 | XP_006720992.1 | 7 - 8 | 1060 | 144 | |
XM_011522614.2 | XP_011520916.1 | 6 - 7 | 904 | 144 | |
XM_011522617.2 | XP_011520919.1 | 4 - 5 | 555 | 144 | |
XM_017023575.1 | XP_016879064.1 | 6 - 7 | 904 | 144 | |
XM_017023576.1 | XP_016879065.1 | 5 - 6 | 670 | 144 | |
XM_017023577.1 | XP_016879066.1 | 7 - 8 | 1335 | 144 | |
XM_017023578.1 | XP_016879067.1 | 5 - 6 | 755 | 144 | |
XM_017023579.1 | XP_016879068.1 | 5 - 6 | 763 | 144 | |
XM_017023580.1 | XP_016879069.1 | 8 - 9 | 1485 | 144 | |
GenBank mRNA | AB075873.1 | - | 6 - 7 | 890 | 144 |
AK022743.1 | - | 6 - 7 | 904 | 144 | |
AK025955.1 | - | 6 - 7 | 838 | 144 | |
AK093050.1 | - | 3 - 4 | 413 | 144 | |
AK125201.1 | - | 7 - 8 | 1037 | 144 | |
AK225065.1 | - | 6 - 7 | 904 | 144 | |
AK225406.1 | - | 6 - 7 | 891 | 144 | |
BC010738.1 | - | 5 - 6 | 671 | 144 |