The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006244.3 | NP_006235.1 | 9 - 10 | 1567 | 78 |
XM_011545132.2 | XP_011543434.1 | 10 - 11 | 1000 | 78 | |
XM_011545133.1 | XP_011543435.1 | 11 - 12 | 1120 | 78 | |
XM_011545134.2 | XP_011543436.1 | 9 - 10 | 2406 | 78 | |
GenBank mRNA | BC045619.1 | - | 9 - 10 | 1561 | 78 |
L42374.1 | - | 9 - 10 | 1270 | 78 |