Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CACNA1D
- Entrez Gene ID
-
776
- Gene Name
- calcium voltage-gated channel subunit alpha1 D
- Gene Aliases
-
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
- Chromosome Location
-
Chr.3: 53495049 - 53813151 on Build GRCh38
View Full Product Details