The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000720.3 | NP_000711.1 | 30 - 31 | 3958 | 80 |
NM_001128839.2 | NP_001122311.1 | 29 - 30 | 3898 | 80 | |
NM_001128840.2 | NP_001122312.1 | 29 - 30 | 3898 | 80 | |
XM_005265448.3 | XP_005265505.1 | 29 - 30 | 3780 | 80 | |
XM_011534094.2 | XP_011532396.1 | 30 - 31 | 3966 | 80 | |
XM_011534096.2 | XP_011532398.1 | 29 - 30 | 3906 | 80 | |
XM_011534097.2 | XP_011532399.1 | 28 - 29 | 4433 | 80 | |
XM_011534099.2 | XP_011532401.1 | 26 - 27 | 3416 | 80 | |
XM_011534100.2 | XP_011532402.1 | 29 - 30 | 3906 | 80 | |
XM_017007137.1 | XP_016862626.1 | 30 - 31 | 3966 | 80 | |
XM_017007138.1 | XP_016862627.1 | 30 - 31 | 3963 | 80 | |
XM_017007139.1 | XP_016862628.1 | 30 - 31 | 3966 | 80 | |
XM_017007140.1 | XP_016862629.1 | 29 - 30 | 3906 | 80 | |
XM_017007141.1 | XP_016862630.1 | 29 - 30 | 3906 | 80 | |
XM_017007142.1 | XP_016862631.1 | 30 - 31 | 3966 | 80 | |
XM_017007145.1 | XP_016862634.1 | 30 - 31 | 3966 | 80 | |
GenBank mRNA | AK294585.1 | - | 4 - 5 | 533 | 80 |
EU363339.1 | - | 29 - 30 | 3780 | 80 | |
M76558.1 | - | 29 - 30 | 4290 | 80 | |
M83566.1 | - | 30 - 31 | 3958 | 80 |