The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001739.1 | NP_001730.1 | 3 - 4 | 522 | 100 |
XM_011523309.2 | XP_011521611.1 | 3 - 4 | 591 | 100 | |
XM_011523310.2 | XP_011521612.1 | 3 - 4 | 591 | 100 | |
XM_017023646.1 | XP_016879135.1 | 3 - 4 | 591 | 100 | |
GenBank mRNA | BC137405.1 | - | 3 - 4 | 518 | 100 |
BC137411.1 | - | 3 - 4 | 518 | 100 | |
L19297.1 | - | 3 - 4 | 521 | 100 |