The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271938.1 | NP_001258867.1 | 20 - 21 | 4180 | 103 |
NM_001410.2 | NP_001401.2 | 19 - 20 | 3979 | 103 | |
GenBank mRNA | AB011541.2 | - | 19 - 20 | 3979 | 103 |
AY280362.1 | - | 14 - 15 | 2302 | 103 | |
BC153880.1 | - | 19 - 20 | 3979 | 103 |