The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135955.1 | NP_001129427.1 | 2 - 3 | 615 | 93 |
NM_001135956.1 | NP_001129428.1 | 2 - 3 | 615 | 93 | |
NM_001135957.1 | NP_001129429.1 | 2 - 3 | 615 | 93 | |
NM_003306.1 | NP_003297.1 | 2 - 3 | 615 | 93 | |
NM_016179.2 | NP_057263.1 | 2 - 3 | 615 | 93 | |
XM_017020724.1 | XP_016876213.1 | 2 - 3 | 1187 | 93 | |
GenBank mRNA | AF063822.1 | - | 2 - 3 | 615 | 93 |
AF063823.1 | - | 2 - 3 | 615 | 93 | |
AF063824.1 | - | 2 - 3 | 615 | 93 | |
AF063825.1 | - | 2 - 3 | 615 | 93 | |
AF175406.1 | - | 2 - 3 | 611 | 93 | |
AF421358.1 | - | 1 - 2 | 380 | 93 | |
AF421359.1 | - | 1 - 2 | 380 | 93 | |
AF421360.1 | - | 1 - 2 | 380 | 93 | |
AF421361.1 | - | 1 - 2 | 380 | 93 | |
AK312524.1 | - | 2 - 3 | 585 | 93 | |
BC104725.1 | - | 2 - 3 | 536 | 93 | |
BC105306.1 | - | 2 - 3 | 536 | 93 | |
BC105641.1 | - | 2 - 3 | 536 | 93 | |
BC105917.1 | - | 2 - 3 | 536 | 93 | |
BX648312.1 | - | 2 - 3 | 1239 | 93 |