Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
Chromosome Location
Chr.22: 30285118 - 30289627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037666.2 NP_001032755.1 3 - 4 504 78
GenBank mRNA AK055587.1 - 3 - 4 410 78
AK309436.1 - 2 - 3 337 78
AK310246.1 - 2 - 3 352 78
BC021927.1 - 3 - 4 390 78
CR456449.1 - 3 - 4 419 78
DA343534.1 - 3 - 4 504 78

View Full Product Details