The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164741.1 | NP_001158213.1 | 20 - 21 | 2590 | 98 |
NM_001666.4 | NP_001657.3 | 19 - 20 | 2470 | 98 | |
GenBank mRNA | AK294562.1 | - | 19 - 20 | 2395 | 98 |
AY183123.1 | - | 16 - 17 | 2036 | 98 | |
BC023626.2 | - | 5 - 6 | 644 | 98 | |
BC052303.1 | - | 20 - 21 | 2590 | 98 | |
D50921.1 | - | 19 - 20 | 2401 | 98 | |
X78817.1 | - | 19 - 20 | 2454 | 98 |