Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
Chromosome Location
Chr.16: 67229389 - 67247522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318202.1 NP_001305131.1 8 - 9 965 75
NM_013241.2 NP_037373.2 8 - 9 965 75
XM_011523043.2 XP_011521345.1 8 - 9 1181 75
XM_011523044.1 XP_011521346.1 8 - 9 910 75
XM_011523045.2 XP_011521347.1 8 - 9 675 75
GenBank mRNA AB209584.1 - 6 - 7 1434 75
AF113615.1 - 8 - 9 871 75
AK304232.1 - 8 - 9 908 75
AY192154.1 - 8 - 9 853 75
BC033084.1 - 8 - 9 910 75
KJ658417.1 - 8 - 9 853 75

View Full Product Details