Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FIGLA
Entrez Gene ID
344018
Gene Name
folliculogenesis specific bHLH transcription factor
Gene Aliases
BHLHC8, FIGALPHA, POF6
Chromosome Location
Chr.2: 70777310 - 70790643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004311.3 NP_001004311.2 2 - 3 391 101

View Full Product Details