The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000396.3 | NP_000387.1 | 1 - 2 | 228 | 88 |
GenBank mRNA | AK130134.1 | - | 1 - 2 | 47 | 88 |
AK308229.1 | - | 1 - 2 | 97 | 88 | |
AK312880.1 | - | 1 - 2 | 45 | 88 | |
AU118043.1 | - | 1 - 2 | 125 | 88 | |
AU119985.1 | - | 1 - 2 | 123 | 88 | |
AY429530.1 | - | 1 - 2 | 37 | 88 | |
BC016058.1 | - | 1 - 2 | 110 | 88 | |
DA251123.1 | - | 1 - 2 | 228 | 88 | |
S79895.1 | - | 1 - 2 | 140 | 88 | |
U13665.1 | - | 1 - 2 | 104 | 88 |