The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323629.1 | NP_001310558.1 | 2 - 3 | 246 | 233 |
NM_001323631.1 | NP_001310560.1 | 2 - 3 | 246 | 233 | |
NM_018097.2 | NP_060567.1 | 2 - 3 | 246 | 144 | |
GenBank mRNA | AK001322.1 | - | 2 - 3 | 204 | 144 |
BC010903.1 | - | 2 - 3 | 225 | 144 | |
DB450370.1 | - | 2 - 3 | 246 | 144 |