Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
23386
Gene Name
NudC domain containing 3
Gene Aliases
NudCL
Chromosome Location
Chr.7: 44382366 - 44490875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015332.3 NP_056147.2 4 - 5 975 54
XM_011515247.2 XP_011513549.1 4 - 5 1064 54
XM_017011909.1 XP_016867398.1 5 - 6 1001 54
XM_017011910.1 XP_016867399.1 5 - 6 859 54
GenBank mRNA AB028991.1 - 4 - 5 769 54
AK024226.1 - 3 - 4 483 54
AK127163.1 - - 376 54
AK307613.1 - 2 - 3 282 54
BC003691.2 - 4 - 5 836 54
BC011673.2 - 3 - 4 595 54
BC017657.1 - 5 - 6 924 54
BC035014.2 - 4 - 5 873 54
EU350932.1 - 4 - 5 793 54

View Full Product Details