The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015332.3 | NP_056147.2 | 5 - 6 | 1160 | 57 |
XM_017011909.1 | XP_016867398.1 | 6 - 7 | 1186 | 57 | |
XM_017011910.1 | XP_016867399.1 | 6 - 7 | 1044 | 57 | |
GenBank mRNA | AB028991.1 | - | 5 - 6 | 954 | 57 |
AK024226.1 | - | 4 - 5 | 668 | 57 | |
AK127163.1 | - | - | 561 | 57 | |
AK307613.1 | - | 3 - 4 | 467 | 57 | |
BC003691.2 | - | 5 - 6 | 1021 | 57 | |
BC011673.2 | - | 4 - 5 | 780 | 57 | |
BC017657.1 | - | 6 - 7 | 1109 | 57 | |
BC035014.2 | - | 5 - 6 | 1058 | 57 | |
EU350932.1 | - | 5 - 6 | 978 | 57 |