Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
Chromosome Location
Chr.22: 30285118 - 30289627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037666.2 NP_001032755.1 2 - 3 315 75
GenBank mRNA AK055587.1 - 2 - 3 221 75
BC021927.1 - 2 - 3 201 75
CR456449.1 - 2 - 3 230 75
DA343534.1 - 2 - 3 315 75

View Full Product Details