Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NDUFA10
- Entrez Gene ID
-
4705
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A10
- Gene Aliases
-
CI-42KD, CI-42k
- Chromosome Location
-
Chr.2: 239892450 - 240025402 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF453834.1 |
- |
5 - 6 |
664 |
83 |
View Full Product Details