Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTLL9
- Entrez Gene ID
-
164395
- Gene Name
- tubulin tyrosine ligase like 9
- Gene Aliases
-
C20orf125
- Chromosome Location
-
Chr.20: 31870702 - 31944959 on Build GRCh38
View Full Product Details