Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA16
Entrez Gene ID
79612
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases
NARG1L
Chromosome Location
Chr.13: 41311205 - 41377031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024561.4 NP_078837.3 18 - 19 2622 68
XM_005266523.2 XP_005266580.1 11 - 12 1474 68
XM_006719866.2 XP_006719929.1 16 - 17 2418 68
XM_017020744.1 XP_016876233.1 14 - 15 1941 68
GenBank mRNA AK025707.1 - 5 - 6 660 68
AK291248.1 - 18 - 19 2536 68
AL833341.1 - 12 - 13 1599 68
BC028112.1 - 8 - 9 1209 68
BC101719.1 - 18 - 19 2394 68
BC101721.1 - 18 - 19 2394 68

View Full Product Details