The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005797.3 | NP_005788.1 | 5 - 6 | 1038 | 152 |
GenBank mRNA | AF030455.1 | - | 5 - 6 | 774 | 152 |
AF275945.1 | - | 5 - 6 | 784 | 152 | |
AF304447.1 | - | 5 - 6 | 779 | 152 | |
AK290326.1 | - | 5 - 6 | 755 | 152 | |
AY359061.1 | - | 5 - 6 | 775 | 152 |