Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
Chromosome Location
Chr.4: 145619388 - 145660035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172250.2 NP_758454.1 4 - 5 818 78
XM_011531684.2 XP_011529986.1 4 - 5 896 78
XM_011531685.2 XP_011529987.1 4 - 5 939 78
XM_011531686.2 XP_011529988.1 3 - 4 1086 78
GenBank mRNA AK126662.1 - 4 - 5 818 78
BC101178.2 - 3 - 4 780 78
BC101179.2 - 3 - 4 780 78
BC101181.2 - 3 - 4 780 78

View Full Product Details