Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KDM1B
Entrez Gene ID
221656
Gene Name
lysine demethylase 1B
Gene Aliases
AOF1, C6orf193, LSD2, bA204B7.3, dJ298J15.2
Chromosome Location
Chr.6: 18155388 - 18223854 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153042.3 NP_694587.3 9 - 10 988 65
XM_005248925.1 XP_005248982.1 11 - 12 1384 65
XM_005248926.1 XP_005248983.1 11 - 12 1384 65
XM_005248928.1 XP_005248985.1 8 - 9 949 65
XM_011514385.1 XP_011512687.1 9 - 10 988 65
XM_011514386.1 XP_011512688.1 9 - 10 988 65
XM_017010440.1 XP_016865929.1 12 - 13 1411 65
XM_017010441.1 XP_016865930.1 12 - 13 1411 65
XM_017010442.1 XP_016865931.1 11 - 12 1257 65
XM_017010443.1 XP_016865932.1 11 - 12 1204 65
XM_017010445.1 XP_016865934.1 10 - 11 1015 65
XM_017010446.1 XP_016865935.1 12 - 13 1411 65
GenBank mRNA AK125318.1 - 9 - 10 924 65

View Full Product Details