The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077653.2 | NP_001071121.1 | 6 - 7 | 1368 | 93 |
XM_017012456.1 | XP_016867945.1 | 4 - 5 | 3333 | 93 | |
GenBank mRNA | DQ986374.1 | - | 6 - 7 | 1415 | 93 |