The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256456.1 | NP_001243385.1 | 15 - 16 | 1953 | 85 |
NM_001256460.1 | NP_001243389.1 | 14 - 15 | 1587 | 85 | |
NM_001256462.1 | NP_001243391.1 | 11 - 12 | 1186 | 85 | |
NM_001256463.1 | NP_001243392.1 | 11 - 12 | 1177 | 85 | |
NM_017871.5 | NP_060341.2 | 13 - 14 | 1480 | 85 | |
XM_011541647.1 | XP_011539949.1 | 15 - 16 | 1631 | 85 | |
XM_011541648.1 | XP_011539950.1 | 15 - 16 | 1635 | 85 | |
XM_011541650.1 | XP_011539952.1 | 13 - 14 | 1360 | 85 | |
XM_017001557.1 | XP_016857046.1 | 14 - 15 | 1436 | 85 | |
XM_017001558.1 | XP_016857047.1 | 15 - 16 | 1490 | 85 | |
GenBank mRNA | AK000549.1 | - | 5 - 6 | 624 | 85 |
AK021939.1 | - | 11 - 12 | 1143 | 85 | |
AK023356.1 | - | 13 - 14 | 1398 | 85 | |
AK056652.1 | - | 15 - 16 | 1875 | 85 | |
AK058018.1 | - | 11 - 12 | 1152 | 85 | |
AK092190.1 | - | 10 - 11 | 1213 | 85 | |
AK123641.1 | - | 3 - 4 | 2353 | 85 | |
AK291387.1 | - | 15 - 16 | 1916 | 85 | |
AK297350.1 | - | 14 - 15 | 1537 | 85 | |
AL136813.1 | - | 13 - 14 | 1459 | 85 | |
BC000675.2 | - | 10 - 11 | 1020 | 85 | |
BC007978.2 | - | 13 - 14 | 1442 | 85 | |
BC013904.2 | - | 9 - 10 | 889 | 85 | |
CR533557.1 | - | 13 - 14 | 1401 | 85 |