The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256456.1 | NP_001243385.1 | 10 - 11 | 1320 | 66 |
NM_001256460.1 | NP_001243389.1 | 9 - 10 | 954 | 66 | |
NM_001256462.1 | NP_001243391.1 | 6 - 7 | 553 | 66 | |
NM_001256463.1 | NP_001243392.1 | 6 - 7 | 544 | 66 | |
NM_017871.5 | NP_060341.2 | 8 - 9 | 847 | 66 | |
XM_011541647.1 | XP_011539949.1 | 10 - 11 | 998 | 66 | |
XM_011541648.1 | XP_011539950.1 | 10 - 11 | 1002 | 66 | |
XM_011541650.1 | XP_011539952.1 | 8 - 9 | 727 | 66 | |
XM_017001557.1 | XP_016857046.1 | 9 - 10 | 803 | 66 | |
XM_017001558.1 | XP_016857047.1 | 10 - 11 | 857 | 66 | |
GenBank mRNA | AK021939.1 | - | 6 - 7 | 66 | |
AK056652.1 | - | 10 - 11 | 1242 | 66 | |
AK058018.1 | - | 6 - 7 | 519 | 66 | |
AK291387.1 | - | 10 - 11 | 1283 | 66 | |
AK297350.1 | - | 9 - 10 | 904 | 66 | |
AL136813.1 | - | 8 - 9 | 826 | 66 | |
BC000675.2 | - | 5 - 6 | 387 | 66 | |
BC007978.2 | - | 8 - 9 | 809 | 66 | |
BC013904.2 | - | 4 - 5 | 256 | 66 | |
CR533557.1 | - | 8 - 9 | 768 | 66 |