The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130043.1 | NP_001123515.1 | 1 - 2 | 453 | 151 |
NM_001889.3 | NP_001880.2 | 1 - 2 | 453 | 151 | |
XM_017000368.1 | XP_016855857.1 | 1 - 2 | 140 | 151 | |
GenBank mRNA | AK128794.1 | - | 1 - 2 | 58 | 151 |
AK223150.1 | - | 1 - 2 | 85 | 151 | |
AK223201.1 | - | 1 - 2 | 85 | 151 | |
AK314813.1 | - | 1 - 2 | 93 | 151 | |
BC039578.1 | - | 1 - 2 | 75 | 151 | |
DA256305.1 | - | 1 - 2 | 137 | 151 |