Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMHR2
Entrez Gene ID
269
Gene Name
anti-Mullerian hormone receptor type 2
Gene Aliases
AMHR, MISR2, MISRII, MRII
Chromosome Location
Chr.12: 53423855 - 53431672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164690.1 NP_001158162.1 1 - 2 127 105
NM_001164691.1 NP_001158163.1 1 - 2 127 105
NM_020547.2 NP_065434.1 1 - 2 127 105
XM_011538173.1 XP_011536475.1 1 - 2 127 105
XM_011538174.1 XP_011536476.1 1 - 2 127 105
XM_011538176.1 XP_011536478.1 1 - 2 127 105
XM_011538178.1 XP_011536480.1 1 - 2 127 105
XM_011538179.1 XP_011536481.1 1 - 2 127 105
XM_011538180.1 XP_011536482.1 1 - 2 127 105
XM_011538181.1 XP_011536483.1 1 - 2 127 105
XM_011538183.2 XP_011536485.1 1 - 2 127 105
XM_011538184.2 XP_011536486.1 1 - 2 127 105
XM_011538185.1 XP_011536487.1 1 - 2 127 105
XM_011538187.2 XP_011536489.1 1 - 2 127 105
XM_017019179.1 XP_016874668.1 1 - 2 127 105
GenBank mRNA AF172932.1 - 1 - 2 110 105
AK313593.1 - 1 - 2 116 105
BC126316.1 - 1 - 2 102 105
BC136356.1 - 1 - 2 102 105
DB020573.1 - 1 - 2 127 105

View Full Product Details